siRNA Details
| Gene Symbol: Cep70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023873.3 | NP_076362.2 | 14 | 1406 | ||
| GenBank mRNA | AF242319.1 | - | 14 | 1442 | ||
| AK133048.1 | - | 14 | 1412 | |||
| BC050819.1 | - | 14 | 1398 | 6517145 | ||
| BC071260.1 | - | 14 | 1406 | 6336351 | ||

Cep70
Genomic Map