siRNA Details
Gene Symbol: Fam120aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014080.1 | - | 2 | 284 | ||
AK021143.1 | - | 2 | 246 | |||
BC024672.1 | - | 379 | ||||
BC045201.1 | - | 2 | 486 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam120aos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014080.1 | - | 2 | 284 | ||
AK021143.1 | - | 2 | 246 | |||
BC024672.1 | - | 379 | ||||
BC045201.1 | - | 2 | 486 |