siRNA Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026573.2 | NP_080849.1 | 9 | 1341 | ||
| GenBank mRNA | AK145431.1 | - | 9 | 1344 | ||
| BC047049.1 | - | 7 | 1000 | |||
| BC117769.1 | - | 4 | 687 | 40091025 | ||
| BC117770.1 | - | 4 | 687 | 40091027 | ||

Upf3b
Genomic Map