siRNA Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026573.2 | NP_080849.1 | 1, 2 | 255 | ||
| GenBank mRNA | AK011606.1 | - | 1, 2 | 253 | ||
| AK019989.1 | - | 1, 2 | 233 | |||
| AK145431.1 | - | 1, 2 | 258 | |||
| AK162222.1 | - | 1, 2 | 251 | |||
| BC058761.1 | - | 1, 2 | 236 | |||
| BC117769.1 | - | 1, 2 | 273 | 40091025 | ||
| BC117770.1 | - | 1, 2 | 273 | 40091027 | ||

Upf3b
Genomic Map