siRNA Details
| Gene Symbol: Etaa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026576.3 | NP_080852.2 | 5 | 1137 | ||
| GenBank mRNA | AK018594.1 | - | 1 | 434 | ||
| AK019965.1 | - | 5 | 1138 | |||
| AK034138.1 | - | 5 | 1130 | |||
| BC028916.1 | - | 5 | 1132 | 3676092 | ||

Etaa1
Genomic Map