siRNA Details
| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026577.3 | NP_080853.3 | 4, 5 | 788 | ||
| GenBank mRNA | AK016794.1 | - | 4, 5 | 734 | ||
| AK021270.1 | - | 4, 5 | 790 | |||
| BC082574.1 | - | 4, 5 | 788 | 30550373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl13b| Gene Symbol: Arl13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026577.3 | NP_080853.3 | 4, 5 | 788 | ||
| GenBank mRNA | AK016794.1 | - | 4, 5 | 734 | ||
| AK021270.1 | - | 4, 5 | 790 | |||
| BC082574.1 | - | 4, 5 | 788 | 30550373 | ||