siRNA Details
| Gene Symbol: Fam212a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026597.3 | NP_080873.2 | 2 | 383 | ||
| GenBank mRNA | AK020090.1 | - | 2 | 347 | ||
| AK076092.1 | - | 2 | 347 | |||
| BC051538.1 | - | 1 | 124 | |||
| BC115538.1 | - | 2 | 322 | 40101503 | ||

Fam212a
Genomic Map