siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB186125.1 | - | 2 | 1940 | ||
| AK040774.1 | - | 2 | 1458 | |||
| AK173252.1 | - | 1 | 481 | |||
| BC139006.1 | - | 1 | 1412 | 8862028 | ||
| BC158061.1 | - | 1 | 1415 | 9007361 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cgnl1| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB186125.1 | - | 2 | 1940 | ||
| AK040774.1 | - | 2 | 1458 | |||
| AK173252.1 | - | 1 | 481 | |||
| BC139006.1 | - | 1 | 1412 | 8862028 | ||
| BC158061.1 | - | 1 | 1415 | 9007361 | ||