siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB186125.1 | - | 2 | 1033 | ||
| AK036871.1 | - | 2 | 598 | |||
| AK040774.1 | - | 2 | 554 | |||
| AK081179.1 | - | 2 | 558 | |||
| BC139006.1 | - | 1 | 508 | 8862028 | ||
| BC158061.1 | - | 1 | 508 | 9007361 | ||

Cgnl1
Genomic Map