siRNA Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB186125.1 | - | 3 | 2212 | ||
| AK036871.1 | - | 3 | 1774 | |||
| AK040774.1 | - | 3 | 1730 | |||
| AK173252.1 | - | 2 | 753 | |||
| BC139006.1 | - | 2 | 1684 | 8862028 | ||
| BC158061.1 | - | 2 | 1687 | 9007361 | ||

Cgnl1
Genomic Map