siRNA Details
| Gene Symbol: Hsbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024219.1 | NP_077181.1 | 2 | 109 | ||
| GenBank mRNA | AK010939.1 | - | 663 | |||
| AK018708.1 | - | 2 | 141 | |||
| AK050550.1 | - | 2 | 159 | |||
| AK078498.1 | - | 2 | 141 | |||
| AK145940.1 | - | 2 | 119 | |||
| BC002153.1 | - | 2 | 109 | 3485301 | ||

Hsbp1
Genomic Map