siRNA Details
| Gene Symbol: Hsbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024219.1 | NP_077181.1 | 4 | 972 | ||
| GenBank mRNA | AK050550.1 | - | 4 | 1022 | ||
| AK078498.1 | - | 4 | 1004 | |||
| AK145940.1 | - | 4 | 982 | |||
| BC002153.1 | - | 4 | 972 | 3485301 | ||

Hsbp1
Genomic Map