siRNA Details
| Gene Symbol: Urm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026615.4 | NP_080891.1 | 5 | 1213 | ||
| GenBank mRNA | AK049849.1 | - | 5 | 1214 | ||
| AK165008.1 | - | 2 | 2213 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urm1| Gene Symbol: Urm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026615.4 | NP_080891.1 | 5 | 1213 | ||
| GenBank mRNA | AK049849.1 | - | 5 | 1214 | ||
| AK165008.1 | - | 2 | 2213 | |||