siRNA Details
| Gene Symbol: Gsto2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026619.2 | NP_080895.2 | 7 | 892 | ||
| NM_030051.1 | NP_084327.1 | 7 | 845 | |||
| GenBank mRNA | AK018953.1 | - | 7 | 884 | ||
| AK019582.1 | - | 7 | 811 | |||
| AK075753.1 | - | 7 | 893 | |||
| AK077086.1 | - | 7 | 846 | |||
| BC030371.1 | - | 5 | 750 | 5371983 | ||

Gsto2
Genomic Map