siRNA Details
| Gene Symbol: Fam24a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183272.2 | NP_899095.1 | 3 | 270 | ||
| GenBank mRNA | AK006815.1 | - | 3 | 271 | ||
| AK018853.1 | - | 3 | 272 | |||
| AK018873.1 | - | 3 | 310 | |||
| AK018966.1 | - | 3 | 310 | |||
| BC048642.1 | - | 3 | 327 | 6772199 | ||

Fam24a
Genomic Map