siRNA Details
| Gene Symbol: AI846148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033139.3 | NP_001028311.2 | 1, 2 | 360 | ||
| NM_001167831.1 | NP_001161303.1 | 1, 2 | 360 | |||
| GenBank mRNA | AK090385.1 | - | 1, 2 | 329 | ||
| AK163590.1 | - | 1, 2 | 161 | |||
| AK166227.1 | - | 1, 2 | 328 | |||
| AK168010.1 | - | 1, 2 | 256 | |||
| BC125249.1 | - | 1, 2 | 188 | 40059530 | ||
| CJ166493.1 | - | 1, 2 | 360 | |||

AI846148
Genomic Map