siRNA Details
| Gene Symbol: H2afb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026627.1 | NP_080903.1 | 1 | 467 | ||
| GenBank mRNA | AK006586.1 | - | 1 | 453 | ||
| AK015538.1 | - | 1 | 458 | |||
| AK018839.1 | - | 1 | 467 | |||
| AK018883.1 | - | 1 | 389 | |||
| AK018922.1 | - | 1 | 453 | |||
| AK018951.1 | - | 1 | 453 | |||
| AK018962.1 | - | 1 | 453 | |||
| AK160415.1 | - | 1 | 453 | |||
| BC048736.1 | - | 1 | 431 | |||
| BC061062.1 | - | 1 | 479 | 6774311 | ||

H2afb1
Genomic Map