siRNA Details
| Gene Symbol: Fam229a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085491.2 | NP_001078960.1 | 3 | 492 | ||
| GenBank mRNA | AK006760.1 | - | 3 | 460 | ||
| AK018907.1 | - | 2 | 521 | |||
| AK018964.1 | - | 3 | 445 | |||
| AK018987.1 | - | 3 | 475 | |||
| BC048660.1 | - | 2 | 489 | |||
| BY707048.1 | - | 2 | 517 | |||
| CA465983.1 | - | 3 | 492 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109266.

Fam229a
Genomic Map