siRNA Details
| Gene Symbol: Fam229a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085491.2 | NP_001078960.1 | 2 | 274 | ||
| GenBank mRNA | AK006760.1 | - | 2 | 242 | ||
| AK018907.1 | - | 1 | 303 | |||
| AK018964.1 | - | 2 | 227 | |||
| AK018987.1 | - | 2 | 257 | |||
| BC048660.1 | - | 1 | 271 | |||
| BC115675.1 | - | 1 | 344 | |||
| BC150491.1 | - | 1 | 344 | |||
| BC150492.1 | - | 1 | 344 | |||
| BY707048.1 | - | 1 | 299 | |||
| CA465983.1 | - | 2 | 274 | |||

Fam229a
Genomic Map