siRNA Details
| Gene Symbol: Mturn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289740.1 | NP_001276669.1 | 2 | 361 | ||
| NM_001289741.1 | NP_001276670.1 | 2 | 361 | |||
| GenBank mRNA | AK019120.1 | - | 2 | 242 | ||
| AK019128.1 | - | 2 | 240 | |||
| AK043452.1 | - | 2 | 214 | |||
| BC042507.1 | - | 2 | 253 | 4235651 | ||
| BC052467.1 | - | 2 | 245 | 6834900 | ||
| BY266709.1 | - | 2 | 361 | |||

Mturn
Genomic Map