siRNA Details
| Gene Symbol: Krt42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212483.2 | NP_997648.2 | 1 | 384 | ||
| GenBank mRNA | AY459292.1 | - | 1 | 384 | ||
| BC112371.1 | - | 1 | 376 | |||
| BC148204.1 | - | 1 | 384 | 40056537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt42| Gene Symbol: Krt42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212483.2 | NP_997648.2 | 1 | 384 | ||
| GenBank mRNA | AY459292.1 | - | 1 | 384 | ||
| BC112371.1 | - | 1 | 376 | |||
| BC148204.1 | - | 1 | 384 | 40056537 | ||