siRNA Details
| Gene Symbol: Krt42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212483.2 | NP_997648.2 | 6 | 1103 | ||
| GenBank mRNA | AK019115.1 | - | 3 | 620 | ||
| AK019133.1 | - | 3 | 565 | |||
| AY459292.1 | - | 6 | 1103 | |||
| BC112371.1 | - | 6 | 1095 | |||
| BC148204.1 | - | 6 | 1103 | 40056537 | ||

Krt42
Genomic Map