siRNA Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026641.2 | NP_080917.1 | 15 | 1828 | ||
| GenBank mRNA | AK019542.1 | - | 2 | 1745 | ||
| AK042231.1 | - | 15 | 1753 | |||
| AK129339.1 | - | 13 | 2076 | |||
| AK147305.1 | - | 14 | 5355 | |||
| BC013814.1 | - | 10 | 1117 | |||
| BC034101.1 | - | 15 | 1723 | 4501451 | ||

Ift80
Genomic Map