siRNA Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026641.2 | NP_080917.1 | 19 | 2379 | ||
| GenBank mRNA | AK019542.1 | - | 6 | 2296 | ||
| AK042231.1 | - | 19 | 2304 | |||
| AK147305.1 | - | 18 | 5906 | |||
| BC013814.1 | - | 14 | 1668 | |||
| BC034101.1 | - | 19 | 2274 | 4501451 | ||

Ift80
Genomic Map