siRNA Details
| Gene Symbol: Zdhhc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026647.3 | NP_080923.2 | 6 | 799 | ||
| GenBank mRNA | AK020293.1 | - | 6 | 681 | ||
| AY166674.1 | - | 4 | 437 | |||
| BC043110.1 | - | 6 | 645 | 6406930 | ||
| BC058372.1 | - | 6 | 667 | |||

Zdhhc21
Genomic Map