siRNA Details
| Gene Symbol: Pomgnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290658.1 | NP_001277587.1 | 2, 3 | 309 | ||
| NM_026651.2 | NP_080927.1 | 3, 4 | 395 | |||
| NM_029786.2 | NP_084062.2 | 3, 4 | 395 | |||
| GenBank mRNA | AB053220.1 | - | 3, 4 | 377 | ||
| AK002638.1 | - | 3, 4 | 387 | |||
| AK015478.1 | - | 2, 3 | 311 | |||
| AK019640.1 | - | 2, 3 | 301 | |||
| BC011201.1 | - | 3, 4 | 318 | 4190368 | ||
| BY732188.1 | - | 3, 4 | 395 | |||

Pomgnt1
Genomic Map