siRNA Details
| Gene Symbol: Toporsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026652.2 | NP_080928.1 | 2 | 1512 | ||
| GenBank mRNA | AK006061.1 | - | 1 | 1022 | ||
| AK015747.1 | - | 2 | 1546 | |||
| AK019748.1 | - | 2 | 1513 | |||
| BC120821.1 | - | 1 | 1381 | 40129744 | ||
| BC120823.1 | - | 1 | 1381 | 40129746 | ||

Toporsl
Genomic Map