siRNA Details
| Gene Symbol: 2310057M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026655.3 | NP_080931.1 | 3 | 455 | ||
| GenBank mRNA | AK019097.2 | - | 3 | 457 | ||
| AK088850.1 | - | 3 | 491 | |||
| AK148008.1 | - | 464 | ||||
| BC044749.2 | - | 3 | 463 | 5102741 | ||
| BC052432.1 | - | 3 | 453 | |||

2310057M21Rik
Genomic Map