siRNA Details
| Gene Symbol: Ddx39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197982.3 | NP_932099.2 | 6 | 806 | ||
| GenBank mRNA | AK011285.1 | - | 5 | 645 | ||
| AK012002.1 | - | 6 | 713 | |||
| AK137114.1 | - | 712 | ||||
| AK145927.1 | - | 6 | 749 | |||
| AK146294.1 | - | 6 | 751 | |||
| AK149475.1 | - | 7 | 771 | |||
| AK150997.1 | - | 6 | 735 | |||
| AK151395.1 | - | 7 | 775 | |||
| AK152394.1 | - | 6 | 716 | |||
| AK162752.1 | - | 1274 | ||||
| AK164289.1 | - | 6 | 676 | |||
| AK167890.1 | - | 6 | 676 | |||
| AK168079.1 | - | 6 | 675 | |||
| AK172448.1 | - | 6 | 735 | |||
| BC020134.1 | - | 6 | 694 | 3969048 | ||

Ddx39
Genomic Map