siRNA Details
| Gene Symbol: Mto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026658.2 | NP_080934.2 | 9, 10 | 1732 | ||
| GenBank mRNA | AF369902.1 | - | 9, 10 | 1722 | ||
| AK019088.1 | - | 9, 10 | 1732 | |||
| AK208615.1 | - | 1, 2 | 143 | |||
| BC063256.1 | - | 10 | 4552 | |||

Mto1
Genomic Map