siRNA Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026660.2 | NP_080936.1 | 7, 8 | 1020 | ||
| GenBank mRNA | AK003763.1 | - | 2, 3 | 288 | ||
| AK018736.1 | - | 7, 8 | 1033 | |||
| AK089055.1 | - | 7, 8 | 1037 | |||
| AK152438.1 | - | 7, 8 | 1006 | |||
| AK177341.1 | - | 26 | ||||
| BC023012.1 | - | 7, 8 | 1020 | 5376230 | ||
| BC064775.1 | - | 7, 8 | 918 | 30023877 | ||

Mfsd10
Genomic Map