siRNA Details
| Gene Symbol: Tsr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163718.1 | NP_001157190.1 | 3 | 499 | ||
| NM_026676.3 | NP_080952.2 | 3 | 520 | |||
| GenBank mRNA | AK002309.1 | - | 3 | 492 | ||
| AK003675.1 | - | 1 | 971 | |||
| AK158743.1 | - | 3 | 521 | |||
| AK169072.1 | - | 2 | 855 | |||
| AK170074.1 | - | 3 | 489 | |||
| BC027751.1 | - | 3 | 590 | 5011894 | ||
| BC038884.1 | - | 1 | 293 | 1516275 | ||
| BC089019.1 | - | 3 | 469 | 30551363 | ||
| CA979987.1 | - | 3 | 448 | |||

Tsr3
Genomic Map