siRNA Details
| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293741.1 | NP_001280670.1 | 3, 4 | 240 | ||
| NM_026677.4 | NP_080953.1 | 3, 4 | 371 | |||
| GenBank mRNA | AB232608.1 | - | 3, 4 | 256 | ||
| AK002303.1 | - | 3, 4 | 338 | |||
| AK080805.1 | - | 3, 4 | 241 | |||
| BB867138.1 | - | 3, 4 | 374 | |||
| BC027214.1 | - | 3, 4 | 364 | 3709140 | ||

Rab13
Genomic Map