siRNA Details
| Gene Symbol: Sdhaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033140.3 | NP_001028312.2 | 1 | 464 | ||
| GenBank mRNA | AK002462.1 | - | 1 | 446 | ||
| AK155437.1 | - | 1 | 428 | |||
| CA451543.1 | - | 1 | 436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sdhaf1| Gene Symbol: Sdhaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033140.3 | NP_001028312.2 | 1 | 464 | ||
| GenBank mRNA | AK002462.1 | - | 1 | 446 | ||
| AK155437.1 | - | 1 | 428 | |||
| CA451543.1 | - | 1 | 436 | |||