siRNA Details
| Gene Symbol: Sdhaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033140.3 | NP_001028312.2 | 1 | 198 | ||
| GenBank mRNA | AA050643.1 | - | 1 | 197 | ||
| AK002462.1 | - | 1 | 181 | |||
| AK155437.1 | - | 1 | 162 | |||
| BC147173.1 | - | 1 | 195 | 9056222 | ||
| BC147174.1 | - | 1 | 195 | 9056227 | ||
| CA451543.1 | - | 1 | 170 | |||

Sdhaf1
Genomic Map