siRNA Details
| Gene Symbol: Ccdc88c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026681.4 | NP_080957.2 | 14, 15 | 1822 | ||
| GenBank mRNA | AK137260.1 | - | 14, 15 | 1758 | ||
| AY339881.1 | - | 14, 15 | 1823 | |||
| BC043115.1 | - | 15, 16 | 1856 | 6408630 | ||
| BC063255.1 | - | 15, 16 | 1856 | 6408630 | ||

Ccdc88c
Genomic Map