siRNA Details
| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178848.3 | NP_849179.1 | 7, 8 | 908 | ||
| GenBank mRNA | AK002609.1 | - | 8, 9 | 937 | ||
| AK005346.1 | - | 7, 8 | 921 | |||
| BC031770.1 | - | 7, 8 | 908 | 4014119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sirt5| Gene Symbol: Sirt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178848.3 | NP_849179.1 | 7, 8 | 908 | ||
| GenBank mRNA | AK002609.1 | - | 8, 9 | 937 | ||
| AK005346.1 | - | 7, 8 | 921 | |||
| BC031770.1 | - | 7, 8 | 908 | 4014119 | ||