siRNA Details
| Gene Symbol: 0610030E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026696.1 | NP_080972.1 | 4 | 802 | ||
| GenBank mRNA | AK002701.1 | - | 4 | 748 | ||
| AK085662.1 | - | 1 | 2013 | |||
| BC064080.1 | - | 4 | 785 | 1246433 | ||
| BC066053.1 | - | 4 | 776 | 6827710 | ||
| BC117811.1 | - | 3 | 829 | |||
| BC117812.1 | - | 4 | 752 | 40092656 | ||

0610030E20Rik
Genomic Map