siRNA Details
| Gene Symbol: Tmem129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026698.2 | NP_080974.2 | 2 | 333 | ||
| GenBank mRNA | AK002836.1 | - | 2 | 382 | ||
| AK172044.1 | - | 2 | 462 | |||
| BC029088.1 | - | 2 | 379 | 3587662 | ||
| BC094042.1 | - | 2 | 333 | 5098723 | ||

Tmem129
Genomic Map