siRNA Details
| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 1, 2 | 166 | ||
| GenBank mRNA | AK016995.1 | - | 1, 2 | 472 | ||
| DT911344.1 | - | 1, 2 | 140 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc163| Gene Symbol: Ccdc163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026714.2 | NP_080990.2 | 1, 2 | 166 | ||
| GenBank mRNA | AK016995.1 | - | 1, 2 | 472 | ||
| DT911344.1 | - | 1, 2 | 140 | |||