siRNA Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290765.1 | NP_001277694.1 | 2 | 315 | ||
| NM_026721.3 | NP_080997.1 | 2 | 315 | |||
| GenBank mRNA | AK035296.1 | - | 2 | 222 | ||
| AK156951.1 | - | 2 | 240 | |||
| BC100463.1 | - | 2 | 283 | 30918840 | ||
| BC125525.1 | - | 2 | 176 | 40130040 | ||
| BC137752.1 | - | 2 | 176 | 8860771 | ||
| BX513015.1 | - | 2 | 316 | |||

Slc39a13
Genomic Map