siRNA Details
| Gene Symbol: Dusp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026725.2 | NP_081001.1 | 2 | 781 | ||
| GenBank mRNA | AB183013.1 | - | 2 | 775 | ||
| AK004912.1 | - | 2 | 672 | |||
| BC069187.1 | - | 2 | 775 | 850475 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp23| Gene Symbol: Dusp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026725.2 | NP_081001.1 | 2 | 781 | ||
| GenBank mRNA | AB183013.1 | - | 2 | 775 | ||
| AK004912.1 | - | 2 | 672 | |||
| BC069187.1 | - | 2 | 775 | 850475 | ||