siRNA Details
| Gene Symbol: Ly6g6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023463.3 | NP_075952.1 | 3 | 303 | ||
| GenBank mRNA | AJ315546.1 | - | 3 | 238 | ||
| AJ315547.1 | - | 2 | 580 | |||
| AK003375.1 | - | 3 | 304 | |||
| BC116365.1 | - | 3 | 269 | 40100048 | ||
| BC116366.1 | - | 3 | 267 | 40100049 | ||

Ly6g6c
Genomic Map