siRNA Details
| Gene Symbol: Ssna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023464.2 | NP_075953.1 | 3 | 353 | ||
| GenBank mRNA | AB041656.1 | - | 3 | 347 | ||
| AK003356.2 | - | 3 | 354 | |||
| AK004482.1 | - | 1 | 232 | |||
| AK088418.1 | - | 3 | 285 | |||
| AK158412.1 | - | 470 | ||||
| AK208309.1 | - | 2 | 45 | |||
| BC023033.1 | - | 3 | 316 | |||
| BC037081.1 | - | 3 | 330 | 5098653 | ||
| BC048360.1 | - | 3 | 331 | 5067090 | ||
| BY703613.1 | - | 3 | 354 | |||

Ssna1
Genomic Map