siRNA Details
| Gene Symbol: Rmnd5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024288.2 | NP_077250.2 | 4 | 960 | ||
| GenBank mRNA | AK148556.1 | - | 3 | 195 | ||
| AK149170.1 | - | 3 | 195 | |||
| AK164384.1 | - | 4 | 961 | |||
| BC050876.1 | - | 4 | 811 | 6512227 | ||
| CF748545.1 | - | 4 | 634 | |||

Rmnd5a
Genomic Map