siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178065.4 | NP_835166.3 | 4, 5 | 875 | ||
| GenBank mRNA | AB093230.1 | - | 3, 4 | 494 | ||
| AK077015.1 | - | 3, 4 | 407 | |||
| AK133332.1 | - | 3, 4 | 409 | |||
| AK147491.1 | - | 4, 5 | 876 | |||
| AK147878.1 | - | 4, 5 | 839 | |||
| BC057351.1 | - | 4, 5 | 813 | |||
| BC060658.1 | - | 3, 4 | 444 | 30355406 | ||
| BC079590.1 | - | 4, 5 | 833 | |||

Arel1
Genomic Map