siRNA Details
| Gene Symbol: Ppfia4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144855.1 | NP_001138327.1 | 24 | 3433 | ||
| GenBank mRNA | AK003571.1 | - | 3 | 291 | ||
| AK078263.1 | - | 1 | 470 | |||
| AK170782.1 | - | 2 | 326 | |||
| AK173053.1 | - | 23 | 2896 | |||
| EU568872.1 | - | 24 | 3433 | |||

Ppfia4
Genomic Map