siRNA Details
| Gene Symbol: Ints1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026748.2 | NP_081024.3 | 5 | 1037 | ||
| GenBank mRNA | AB257854.1 | - | 5 | 997 | ||
| AK155838.1 | - | 5 | 1033 | |||
| BC063266.1 | - | 5 | 1012 | 6409930 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ints1| Gene Symbol: Ints1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026748.2 | NP_081024.3 | 5 | 1037 | ||
| GenBank mRNA | AB257854.1 | - | 5 | 997 | ||
| AK155838.1 | - | 5 | 1033 | |||
| BC063266.1 | - | 5 | 1012 | 6409930 | ||