siRNA Details
| Gene Symbol: Eml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043335.1 | NP_001036800.1 | 11 | 1589 | ||
| NM_001043336.2 | NP_001036801.1 | 11 | 1206 | |||
| NM_001286346.1 | NP_001273275.1 | 11 | 1343 | |||
| NM_001286347.1 | NP_001273276.1 | 12 | 1257 | |||
| GenBank mRNA | AK044662.1 | - | 11 | 1328 | ||
| BC053094.1 | - | 11 | 1423 | 6400458 | ||
| BC059839.1 | - | 11 | 1184 | 6417403 | ||
| BC079582.1 | - | 11 | 1242 | 5702743 | ||
| BC098329.1 | - | 10 | 986 | |||
| BC125289.1 | - | 12 | 1228 | 40129804 | ||
| BC144757.1 | - | 12 | 1228 | |||
| BC150975.1 | - | 11 | 1343 | 9087888 | ||

Eml1
Genomic Map