siRNA Details
| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014995.1 | NP_001014995.1 | 7 | 1333 | ||
| GenBank mRNA | BC085505.1 | - | 7 | 1075 | ||
| BC088990.1 | - | 7 | 1333 | 6402466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam189b| Gene Symbol: Fam189b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014995.1 | NP_001014995.1 | 7 | 1333 | ||
| GenBank mRNA | BC085505.1 | - | 7 | 1075 | ||
| BC088990.1 | - | 7 | 1333 | 6402466 | ||